missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Small capsomere-interacting Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Small capsomere-interacting protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7057-500ug
Additional Details : Weight : 0.01000kg
Specifications
YP_401651.1 | |
Small capsomere-interacting Protein | |
Research Use Only | |
BFRF3 | |
Recombinant Protein | |
E. coli | |
NQNNLPNDVF REAQRSYLVF LTSQFCYEEY VQRTFGVPRR QRAIDKRQRA SVAGAGAHAH LGGSSATPVQ QAQAAASAGT GALASSAPST AVAQSATPSV SSSISSLRAA TSGATAAASA AAAVDTGSGG GGQPHDTAPR GARKKQ | |
Greater than 90% as determined by SDS-PAGE. |
3783701 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |