missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Small capsomere-interacting Protein

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Small capsomere-interacting protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7057-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067250

  • 605.00 € / 50µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

YP_401651.1
Small capsomere-interacting Protein
Research Use Only
BFRF3
Recombinant Protein
E. coli
NQNNLPNDVF REAQRSYLVF LTSQFCYEEY VQRTFGVPRR QRAIDKRQRA SVAGAGAHAH LGGSSATPVQ QAQAAASAGT GALASSAPST AVAQSATPSV SSSISSLRAA TSGATAAASA AAAVDTGSGG GGQPHDTAPR GARKKQ
Greater than 90% as determined by SDS-PAGE.
3783701
50 μg
Not determined.
Partial (see sequence information for more details).
Epstein-Barr
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers