missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant F. meningosepticum PNGase F Protein

Product Code. 18757267
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757267 100 μg 100µg
18735817 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757267 Supplier R&D Systems™ Supplier No. 9109GH100
  • ONLINE EXCLUSIVE PRICE
    994.46 €
    892.50 € / 100µg
    Save 101.96 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Effective removal of N-linked oligosaccharides from glycoproteins. Glycerol-free, suitable for downstream MS and HPLC.

Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10 μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21163
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Quantity 100 μg
Source E. coli-derived f. meningosepticum PNGase F protein Ala41-Asn354 with N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Common Name PNGase F
Biological Activity Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Conjugate Unconjugated
Species F. meningosepticum
Recombinant Recombinant
Expression System E. coli-derived Ala41-Asn354 with N-terminal Met and 6-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PNGase F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.