missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori Vacuolating cytotoxin autotransporter Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Vacuolating cytotoxin autotransporter, was expressed in the hosts and tags indicated. option, abo
309.00 € - 3260.00 €
Specifications
Accession Number | NP_207680.1 |
---|---|
Gene ID (Entrez) | 899415 |
Name | Vacuolating cytotoxin autotransporter Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026521
|
enQuireBio™
QP8796-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046521
|
enQuireBio™
QP8796-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036521
|
enQuireBio™
QP8796-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056521
|
enQuireBio™
QP8796-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066521
|
enQuireBio™
QP8796-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076521
|
enQuireBio™
QP8796-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_207680.1 | |
Vacuolating cytotoxin autotransporter Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |
899415 | |
Research Use Only | |
vacA | |
Recombinant Protein | |
E. coli | |
TTVIIPAIVG GIATGAAVGT VSGLLGWGLK QAEEANKTPD KPDKVWRIQA GKGFNEFPNK EYDLYRSLLS SKIDGGWDWG NAATHYWVKG GQWNKLEVDM KDAVGTYNLS GLRNFTGGDL DVNMQKATLR LGQFNGNSFT SYKDSADRTT RVDFNAKNIL IDNFLEINNR VGSGAGRKAS STVLTLQASE GITSSKNAEI SLYDGATLN | |
Greater than 90% as determined by SDS-PAGE. |