missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adw Protein
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
Specifications
Specifications
| Name | Hepatitis B HBsAg adw Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Hepatitis B |
| Species | P. pastoris |
| Protein Tag | Untagged |
| Buffer | Sterile Filtered solution containing 20mM Phosphate Buffer, 154mM sodium chloride, pH 7.1. |
| Purity or Quality Grade | Greater than 95.0% as determined by SDS-PAGE and HPLC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction