missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV Pre-S Protein
A cDNA sequence encoding the HBV Pre-S was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12133-1mg
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis B HBV Pre-S Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing PBS, 1M urea and 0.05% sodium azide. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |