missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-1 Protein

A cDNA sequence encoding the HCV Core Genotype-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12144-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959508

  • 1450.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV Core Genotype-1 Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris pH 8 and 5mM EDTA.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV Core Genotype-1 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers