missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-6a Protein
A cDNA sequence encoding the HCV Core Genotype-6a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12155-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV Core Genotype-6a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione, 0.25% sarkosil and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
HCV Core Genotype-6a protein is >95% pure as determined by 10% PAGE (coomassie staining). |