missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-1b Protein
A cDNA sequence encoding the HCV NS3 Genotype-1b was constructed and used to recombinantly synthesize the protein.
228.00 € - 1450.00 €
Specifications
Name | Hepatitis C HCV NS3 Genotype-1b Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979568
|
enQuireBio™
QP12165-HIS-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999568
|
enQuireBio™
QP12165-HIS-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989568
|
enQuireBio™
QP12165-HIS-1MG |
1 mg |
1450.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis C HCV NS3 Genotype-1b Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His | |
50mM NaPO4 pH 8.3 and 10mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Starts: MRDSDSQTFQ Ends: DSVIDCNTCVT | |
HCV NS3 Genotype-1b protein is >95% pure as determined by 10% PAGE (coomassie staining). |