missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3a Protein
A cDNA sequence encoding the HCV NS5 Genotype-3a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12193-500ug
Weitere Details : Weight : 0.01000kg
Spezifikation
Hepatitis C HCV NS5 Genotype-3a Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25 mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV NS5 Genotype-3a protein is >95% pure as determined by 10% PAGE (coomassie staining). |