missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3a Protein

A cDNA sequence encoding the HCV NS5 Genotype-3a was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12193-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989648

  • 228.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV NS5 Genotype-3a Protein
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25 mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV NS5 Genotype-3a protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers