missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis Capsid Protein
A DNA sequence encoding the Hepatitis E virus genotype 3 (isolate Human/United States/US2) (HEV-3) Capsid protein, was expressed in the hosts and tags indicated. option, abov
352.00 € - 3175.00 €
Specifications
Name | hepatitis Capsid Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | ORF2 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033841
|
enQuireBio™
QP8294-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043841
|
enQuireBio™
QP8294-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023841
|
enQuireBio™
QP8294-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003841
|
enQuireBio™
QP8294-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013841
|
enQuireBio™
QP8294-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
hepatitis Capsid Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
ORF2 | |
Recombinant Protein | |
Yeast | |
KLYTSVENAQ QDKGITIPHD IDLGDSRVVI QDYDNQHEQD RPTPSPAPSR PFSVLRANDV LWLSLTAAEY DQTTYGSSTN PMYVSDTVTL VNVATGAMS- QAVARSLDWS KVTLDGRPLT TIQQYSKTFY VLPLRGKLSF WEAGTTKAGY PYNYNTTASD QILIENAAGH RVAISTYTTS LGAGPTSISA VGVLAPHSAL AVLEDTIDYP ARAHTFDDFC PECRTLGLQG CAFQSTIAEL QRLKMKVGKT RES | |
Greater than 80% as determined by SDS-PAGE. |