missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis E HEV ORF3 Protein
A cDNA sequence encoding the HEV ORF3 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | Hepatitis E HEV ORF3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis E |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919748
|
enQuireBio™
QP12223-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939748
|
enQuireBio™
QP12223-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929748
|
enQuireBio™
QP12223-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis E HEV ORF3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis E | |
Untagged | |
HEV ORF3 protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-HCl, pH 8.0, 10mM B-ME and 8M urea. |