missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant hepatitis protein 1

A DNA sequence encoding the Hepatitis B virus genotype D subtype ayw (isolate France/Tiollais/1979) (HBV-D) Protein X, was expressed in the hosts and tags indicated. opt

Brand:  enQuireBio™ QP7274-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037950

  • 3260.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

hepatitis protein 1
Research Use Only
X
Recombinant Protein
E. coli
MAARLCCQLD PARDVLCLRP VGAESRGRPF SGSLGTLSSP SPSAVPTDHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQILP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCAPAPCNF FTSA
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Hepatitis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers