missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 gp120 CM Protein

A cDNA sequence encoding the HIV-1 gp120 CM was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12244-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989808

  • 288.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 gp120 CM Protein
Research Use Only
Recombinant Protein
Baculovirus
10mM Tris-Cl, pH 7.6, 150mM NaCl and 0.01% Triton N-101.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers