missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp120 MN Protein
A cDNA sequence encoding the HIV-1 gp120 MN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12246-100ug
This item is not returnable.
View return policy
Specifications
HIV-1 gp120 MN Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The HIV-1 gp120 protein solution contains 10mM Tris-Cl, pH 7.6, 150mM NaCl and 0.01% Triton N-101. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |