missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp41/120 Protein
A cDNA sequence encoding the HIV-1 gp41/120 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12251-500ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 gp41/120 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
150mM NaCl, 25mM sodium phosphate pH 8.0, 50% glycerol and 150mM imidazole. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |