missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 gp41/120 Protein

A cDNA sequence encoding the HIV-1 gp41/120 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12251-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989828

  • 228.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 gp41/120 Protein
Research Use Only
Recombinant Protein
E. coli
150mM NaCl, 25mM sodium phosphate pH 8.0, 50% glycerol and 150mM imidazole.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
His
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers