missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp41 16kDa Protein
A cDNA sequence encoding the HIV-1 gp41 16kDa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12249-100ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 gp41 16kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized from 1 mg/ml in 20mM Na-carbonate, pH 9.6. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |