missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 Protease Protein

Product Code. 15959858
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15959858

Brand: enQuireBio™ QP1226010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the HIV-1 Protease was constructed and used to recombinantly synthesize the protein.

Specifications

Name HIV-1 Protease Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity HIV
Species E. coli
Protein Tag Untagged
Sequence PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF
Buffer The HIV-1 Protease filtered (0.4microm) solution is formulated in 20mM Tris, 20mM MES, 200mM NaCl, 1mM EDTA, 10% (v/v) glycerol and 0.05% 2-mercaptoethanol, pH 6.5.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.