missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-2 gp36 Protein

A cDNA sequence encoding the HIV-2 gp36 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12269-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929888

  • 131.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-2 gp36 Protein
Research Use Only
Recombinant Protein
HIV-2
(1 mg/1ml) in H2O.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 95.0% as determined by HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers