missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 gp36 Protein
A cDNA sequence encoding the HIV-2 gp36 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12269-1mg
Additional Details : Weight : 0.01000kg
Specifications
HIV-2 gp36 Protein | |
Research Use Only | |
Recombinant Protein | |
HIV-2 | |
(1 mg/1ml) in H2O. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC. |