missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV HTLV-1 Envelope Protein

A cDNA sequence encoding the HTLV-1 Envelope was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12353-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930149

  • 228.00 € / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV HTLV-1 Envelope Protein
Research Use Only
Recombinant Protein
E. coli
10mM Na-PO4 pH 6.0, 0.1% SDS and 1mM DTT.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
HTLV-1 Envelope protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers