missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Horse Major allergen Equ c 1 Protein

A DNA sequence encoding the Equus caballus (Horse) Major allergen Equ c 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7892-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031431

  • 940.00 € / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001075966.1
Major allergen Equ c 1 Protein
Research Use Only
LOC100034197
Recombinant Protein
E. coli
QQEENSDVAI RNFDISKISG EWYSIFLASD VKEKIEENGS MRVFVDVIRA LDNSSLYAEY QTKVNGECTE FPMVFDKTEE DGVYSLNYDG YNVFRISEFE NDEHIILYLV NFDKDRPFQL FEFYAREPDV SPEIKEEFVK IVQKRGIVKE NIIDLTKIDR CFQLRGNGVA QA
Greater than 90% as determined by SDS-PAGE.
100034197
100 μg
Not determined.
Full Length
Horse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers