missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse PMP2 / FABP8 Protein
A DNA sequence encoding the Equus caballus (Horse) PMP2 / FABP8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7014-1mg
This item is not returnable.
View return policy
Specifications
PMP2 / FABP8 Protein | |
Research Use Only | |
PMP2 | |
Recombinant Protein | |
E. coli | |
SNKFLGTWKL TSSENFDEYM KALGVGLGTR SLGNLAGPTV IISKSGDVIT IRTESGFKNT EISFKLGQEF EETTADNRKT KSTVTLAGGK LNQVQKWNGN ETTIKRELVD GKMVVECSMA SVVCTRIYEQ V | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Horse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |