missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HPV HPV 18 Protein
A cDNA sequence encoding the HPV 18 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12308-500ug
Additional Details : Weight : 0.01000kg
Specifications
HPV HPV 18 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
VDVYLPPPSVARVVNTDDYVTPTSIFYHAGSSRLLTVGNPYFRVPAGGGNKQDIPKVSAYQYRVFRVQLPDPNKFGLPDTSIYNPETQRLVWACAGVEIGRGQPLGVGLSGHPFYNKLDDTESSHAATSNVSEDVRDNVSVDYKQTQLCILGCAPAIGEHWAKGTACKSRPLSQGDCPPLELKNTVLEDGDMVDTGYGAMDFSTLQDTKCEVPLDICQSICKYPDYLQMSADPYGDSMFFCLRREQLFARHFWNRAGTMGDTVPQSLYIKGTGMPASPGSCVYSPSPSGSIVTSDSQLFNKPYWLHKAQGHNNGVCWHNQLFVTVVDTTPSTNLTICASTQSPVPGQYDATKFKQYSRHVEEYDLQFIFQLCTITLTADVMSYIHSMNSSILEDWNFGVPPPPTTSLVDTYRFVQSVAITCQKDAAPAENKDPYDKLKFWNVDLKEKFSLDLDQYPLGRKFLVQAGLRRKPTIGPRKRSAPSATTSSKPA | |
Protein is >90% pure as determined by 10% PAGE (Coomassie staining). |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HPV | |
GST | |
Recombinant HPV-18 solution in PBS, 3M Urea and 0.02% sodium azide as preservative. |