missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HSV-2 VP13/14 Protein

A cDNA sequence encoding the HSV-2 VP13/14 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12347-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960129

  • 1740.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSV HSV-2 VP13/14 Protein
Research Use Only
Recombinant Protein
E. coli
10 mM Phosphate buffer pH 7.6; 75 mM NaCl.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
His
Protein is >90% pure as determined by SDS PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers