missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-2 VP13/14 Protein
A cDNA sequence encoding the HSV-2 VP13/14 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12347-500ug
Additional Details : Weight : 0.01000kg
Specifications
HSV HSV-2 VP13/14 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10 mM Phosphate buffer pH 7.6; 75 mM NaCl. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
Protein is >90% pure as determined by SDS PAGE. |