missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 14-3-3 sigma / Stratifin / YWHAS Protein
A DNA sequence encoding the Homo sapiens (Human) 14-3-3 sigma / Stratifin / YWHAS, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP872-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006133.1 | |
14-3-3 sigma / Stratifin / YWHAS Protein | |
Research Use Only | |
SFN | |
Recombinant Protein | |
E. coli | |
MERASLIQKA KLAEQAERYE DMAAFMKGAV EKGEELSCEE RNLLSVAYKN VVGGQRAAWR VLSSIEQKSN EEGSEEKGPE VREYREKVET ELQGVCDTVL GLLDSHLIKE AGDAESRVFY LKMKGDYYRY LAEVATGDDK KRIIDSARSA YQEAMDISKK EMPPTNPIRL GLALNFSVFH YEIANSPEEA ISLAKTTFDE AMADLHTLSE DSYKDSTLIM QLLRDNLTLW TADNAGEEGG EAPQEPQS | |
Greater than 90% as determined by SDS-PAGE. |
2810 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |