missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 14-3-3 tau / 14-3-3 theta / YWHAQ Protein
A DNA sequence encoding the Homo sapiens (Human) 14-3-3 tau / 14-3-3 theta / YWHAQ, was expressed in the hosts and tags indicated.
228.00 € - 2440.00 €
Specifications
Accession Number | NP_006817.1 |
---|---|
Gene ID (Entrez) | 10971 |
Name | 14-3-3 tau / 14-3-3 theta / YWHAQ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016181
|
enQuireBio™
QP873-10UG |
10 μg |
228.00 €
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056181
|
enQuireBio™
QP873-50UG |
50 μg |
324.00 €
50µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006181
|
enQuireBio™
QP873-100UG |
100 μg |
555.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036181
|
enQuireBio™
QP873-200UG |
200 μg |
930.00 €
200µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046181
|
enQuireBio™
QP873-500UG |
500 μg |
1535.00 €
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026181
|
enQuireBio™
QP873-1MG |
1 mg |
2440.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_006817.1 | |
14-3-3 tau / 14-3-3 theta / YWHAQ Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10971 | |
Research Use Only | |
YWHAQ | |
Recombinant Protein | |
E. coli | |
MEKTELIQKA KLAEQAERYD DMATCMKAVT EQGAELSNEE RNLLSVAYKN VVGGRRSAWR VISSIEQKTD TSDKKLQLIK DYREKVESEL RSICTTVLEL LDKYLIANAT NPESKVFYLK MKGDYFRYLA EVACGDDRKQ TIDNSQGAYQ EAFDISKKEM QPTHPIRLGL ALNFSVFYYE ILNNPELACT LAKTAFDEAI AELDTLNEDS YKDSTLIMQL LRDNLTLWTS DSAGEECDAA EGAEN | |
Greater than 80% as determined by SDS-PAGE. |