missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 2-5A-dependent ribonuclease Protein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1a (isolate 1) polyprotein, was expressed in the hosts and tags indicated.
131.00 € - 2345.00 €
Specifications
Accession Number | NP_066956.1 |
---|---|
Gene ID (Entrez) | 6041 |
Name | 2-5A-dependent ribonuclease Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026960
|
enQuireBio™
QP6997-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066960
|
enQuireBio™
QP6997-50UG |
50 μg |
324.00 €
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016960
|
enQuireBio™
QP6997-100UG |
100 μg |
520.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046960
|
enQuireBio™
QP6997-200UG |
200 μg |
835.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056960
|
enQuireBio™
QP6997-500UG |
500 μg |
1440.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036960
|
enQuireBio™
QP6997-1MG |
1 mg |
2345.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_066956.1 | |
2-5A-dependent ribonuclease Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6041 | |
Research Use Only | |
RNASEL | |
Recombinant Protein | |
E. coli | |
HPPAEDWKPQ SSHWGAALKD LHRIYRPMIG KLKFFIDEKY KIADTSEGGI YLGFYEKQEV AVKTFCEGSP RAQREVSCLQ SSRENSHLVT FYGSESHRGH LFVCVTLCEQ TLEACLDVHR GEDVENEEDE FARNVLSSIF KAVQELHLSC GYTHQDLQPQ NILIDSKKAA HLADFDKSIK WAGDPQEVKR DLEDLGRLVL YVVKKGSISF EDLKAQSNEE VVQLSPDEET KDLIHRLFHP GEHVRDCLSD LLGHPFFWTW ESRYRTLRNV GNESDIKTRK SESEILRLLQ PGPSEHSKSF DKWTTKINEC VMKKMNKFYE KRGNFYQNTV GDLLKFIRNL GEHIDEEKHK KMKLKIGDPS LYFQKTFPDL VIYVYTKLQN TEYRKHFPQT HSPNKPQCDG AGGASGLASP GC | |
Greater than 90% as determined by SDS-PAGE. |