missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 2-oxoglutarate dehydrogenase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 2-oxoglutarate dehydrogenase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6447-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003941.1 | |
2-oxoglutarate dehydrogenase, mitochondrial Protein | |
Research Use Only | |
OGDH | |
Recombinant Protein | |
E. coli | |
SAPVAAEPFL SGTSSNYVEE MYCAWLENPK SVHKSWDIFF RNTNAGAPPG TAYQSPLPLS RGSLAAVAHA QSLVEAQPNV DKLVEDHLAV QSLIRAYQIR GHHVAQLDPL GILDADLDSS VPADIISSTD KLGFYGLDES DLDKVFHLPT TTFIGGQESA LPLREIIRRL EMAYCQHIGV EFMFINDLEQ CQWIRQKFET PGIMQFTNEE KRTLLARLVR STRFEEFLQR KWSSEKRFGL EGCEVLIPAL KTIIDKSSEN GVDYVIMGMP HRGRLNVLAN VIRKELEQIF CQFDSKLEAA DEGSGDVKYH LGMYHRRINR VTDRNITLSL VANPSHLEAA DPVVMGKTKA EQFYCGDTEG KKVRPRERRA RQIVKAPCSS MEF | |
Greater than 90% as determined by SDS-PAGE. |
4967 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |