missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 39S ribosomal protein L55, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L55, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6384-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042920

  • 2365.00 € / 1mg
Estimated Shipment: 02-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001308213.1
39S ribosomal protein L55, mitochondrial Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
128308
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MRPL55
Recombinant Protein
E. coli
DSSRASLTRV HRQAYARLYP VLLVKQDGST IHIRYREPRR MLAMPIDLDT LSPEERRARL RKREAQLQSR KEYEQELSDD LHVERYRQFW TRTKK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers