missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 4-hydroxyphenylpyruvate dioxygenase Protein
A DNA sequence encoding the Homo sapiens (Human) 4-hydroxyphenylpyruvate dioxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6179-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001165464.1 | |
4-hydroxyphenylpyruvate dioxygenase Protein | |
Research Use Only | |
HPD | |
Recombinant Protein | |
E. coli | |
TTYSDKGAKP ERGRFLHFHS VTFWVGNAKQ AASFYCSKMG FEPLAYRGLE TGSREVVSHV IKQGKIVFVL SSALNPWNKE MGDHLVKHGD GVKDIAFEVE DCDYIVQKAR ERGAKIMREP WVEQDKFGKV KFAVLQTYGD TTHTLVEKMN YIGQFLPGYE APAFMDPLLP KLPKCSLEMI DHIVGNQPDQ EMVSASEWYL KNLQFHRFWS VDDTQVHTEY SSLRSIVVAN YEESIKMPIN EPAPGKKKSQ IQEYVDYNGG AGVQHIALKT EDIITAIRHL RERGLEFLSV PSTYYKQLRE KLKTAKIKVK ENIDALEELK ILVDYDEKGY LLQIFTKPVQ DRPTLFLEVI QRHNHQGFGA GNFNSLFKAF EEEQNLRGNL TNMETNGVVP GM | |
Greater than 90% as determined by SDS-PAGE. |
3242 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |