missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L14 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8323-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034011

  • 505.00 € / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001030168.1
60S ribosomal protein L14 Protein
Research Use Only
RPL14
Recombinant Protein
E. coli
VFRRFVEVGR VAYVSFGPHA GKLVAIVDVI DQNRALVDGP CTQVRRQAMP FKCMQLTDFI LKFPHSAHQK YVRQAWQKAD INTKWAATRW AKKIEARERK AKMTDFDRFK VMKAKKMRNR IIKNEVKKLQ KAALLKASPK KAPGTKGTAA AAAAAAAAKV PAKKITAASK KAPAQKVPAQ KATGQKAAPA PKAQKGQKAP AQKAPAPKAS GKK
Greater than 90% as determined by SDS-PAGE.
9045
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers