missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human A-kinase anchor protein 1

A DNA sequence encoding the Homo sapiens (Human) A-kinase anchor protein 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7671-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050101

  • 735.00 € / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003877.2
A-kinase anchor protein 1
Research Use Only
AKAP4
Recombinant Protein
E. coli
QSPSAPPAKP PSTQRAVISP DGECSIDDLS FYVNRLSSLV IQMAHKEIKE KLEGKSKCLH HSICPSPGNK ERISPRTPAS KIASEMAYEA VELTAAEMRG TGEESREGGQ KSFLYSELSN KSKSGDKQMS QRESKEFADS ISKGLMVYAN QVASDMMVSL MKTLKVHSSG KPIPASVVLK RVLLRHTKEI VSDLIDSCMK NLHNITGVLM TDSDFVSAVK RNLFNQWKQN ATDIMEAMLK RLVSALIGEE KETKSQSLSY ASLKAGSHDP KCRNQSLEFS TMKAEMKERD KGKMKSDPCK SLTSAEKVGE HILKEGLTIW NQKQGNSCKV ATKACSNKDE KGEKINASTD SLAKDLIVSA LKLIQYHLTQ QTKGKDTCEE DCPGSTMGYM AQSTQYEKCG GGQSAKALSV KQLESHRAPG PSTCQKENQH LDSQKMDMSN IVLMLIQKLL NENPFKCEDP CEGENKCSEP RASKAASMSN RSDKAEEQCQ EHQELDCTSG MKQANGQFID KLVESVMKLC LIMAKYSNDG AALAELEEQA ASANKPNFRG TRCIHSGAMP QNYQDSLGHE VIVNNQCSTN SLQKQLQAVL QWIAASQFNV PMLYFMGDKD GQLEKLPQVS AKAAEKGYSV GGLLQEVMKF AKERQPDEAV GKVARKQLLD WLLANL
Greater than 80% as determined by SDS-PAGE.
8852
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers