missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AADAT Protein
A DNA sequence encoding the Homo sapiens (Human) AADAT, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001273611.1 |
---|---|
Gene ID (Entrez) | 51166 |
Name | AADAT Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051591
|
enQuireBio™
QP7920-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071591
|
enQuireBio™
QP7920-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061591
|
enQuireBio™
QP7920-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081591
|
enQuireBio™
QP7920-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091591
|
enQuireBio™
QP7920-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001601
|
enQuireBio™
QP7920-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001273611.1 | |
AADAT Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
51166 | |
Research Use Only | |
AADAT | |
Recombinant Protein | |
E. coli | |
PKSMISLAGG LPNPNMFPFK TAVITVENGK TIQFGEEMMK RALQYSPSAG IPELLSWLKQ LQIKLHNPPT IHYPPSQGQM DLCVTSGSQQ GLCKVFEMII NPGDNVLLDE PAYSGTLQSL HPLGCNIINV ASDESGIVPD SLRDILSRWK PEDAKNPQKN TPKFLYTVPN GNNPTGNSLT SERKKEIYEL ARKYDFLIIE DDPYYFLQFN KFRVPTFLSM DVDGRVIRAD SFSKIISSGL RIGFLTGPKP LIERVILHIQ VSTLHPSTFN QLMISQLLHE WGEEGFMAHV DRVIDFYSNQ KDAILAAADK WLTGLAEWHV PAAGMFLWIK VKGINDVKEL IEEKAVKMGV LMLPGNAFYV DSSAPSPYLR ASFSSASPEQ MDVAFQVLAQ LIKESL | |
Greater than 90% as determined by SDS-PAGE. |