missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active p38 alpha Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The activity of p38 alpha is typically 148-201 nmol/min/mg using an ATF2 substrate (see activity assay protocol).
Specifications
Specifications
| Components | Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol. |
| Purity | >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane) |
| For Use With (Application) | Bioactivity |
| Storage Buffer | Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol. |
| Content And Storage | This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.) |
| Accession Number | NM_139012 |
| Gene ID (Entrez) | 1432 |
| Gene Symbol | MAPK14 |
| Source | Spodoptera frugiperda,Sf 9 (baculovirus)-derived human p38 alpha protein Accession &Num; NM_139012 |
| Quantity | 10 μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction