missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Active ROCK1 Protein

Product Code. 18349667
Click to view available options
:
10ug, Carrier Free
This item is not returnable. View return policy

Product Code. 18349667

Brand: R&D Systems™ 4590KS010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Active ROCK1 Protein is derived from Sf 9 (baculovirus). The Recombinant Human Active ROCK1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol.
Molecular Weight (g/mol) Observed MW: 85kDa
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human ROCK1 protein aa 17-535 Accession &Num; NM_005406
Storage Requirements This product is stable at ≤ -70° C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.
Conjugate Unconjugated
Purity or Quality Grade >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.