missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Activin A / INHBA Protein

A DNA sequence encoding the Homo sapiens (Human) Activin A / INHBA Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6232-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031790

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002183.1
Activin A / INHBA Protein
Research Use Only
INHBA
Recombinant Protein
E. coli
GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Greater than 90% as determined by SDS-PAGE.
3624
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers