missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Activin AB Protein, CF

Product Code. 30114527
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30114527 25 μg 25µg
30114931 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30114527 Supplier R&D Systems™ Supplier No. 1066AB025/CF
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002183 ( beta A) & NP_002184 ( beta B)
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Molecular Weight (g/mol) M.W.-observed: 14 kDa, reducing conditions, M.W.-theroretical: 12.8 kDa ( beta A subunit) and 13 kDa ( beta B subunit)
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Activin AB proteinHuman Activin beta A (Gly311 - Ser426) (N-terminus) Accession # NP_002183.1 Human Activin beta B (Gly293 - Ala407) (N-terminus) Accession # NP_002184.2 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in 4 mM HCl.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Activin AB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.