missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACVRL1 Protein
A cDNA sequence encoding the ACVRL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10956-2ug
Additional Details : Weight : 0.01000kg
Specifications
ACVRL1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
DPVKPSRGPL VTCTCESPHC KGPTCRGAWC TVVLVREEGR HPQEHRGCGN LHRELCRGRP TEFVNHYCCD SHLCNHNVSL VLEATQPPSEQPGTDGQHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACVRL1 protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |