missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADAT2 Protein

A cDNA sequence encoding the ADAT2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10964-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955708

  • 203.00 € / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

134637
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEAKAAPKPA ASGACSVSAE ETEKWMEEAM HMAKEALENT EVPVGCLMVY NNEVVGKGRN EVNQTKNATR HAEMVAIDQV LDWCRQSGKS PSEVFEHTVL YVTVEPCIMC AAALRLMKIP LVVYGCQNER FGGCGSVLNI ASADLPNTGR PFQCIPGYRA EEAVEMLKTF YKQENPNAPK SKVRKKECQK S
Greater than 90% as determined by Analysis by SDS-PAGE.
ADAT2 Protein
Research Use Only
ADAT2
Human
His
The ADAT2 (0.5 mg/ml) protein contains 20mM Tris-HCl buffer pH 8.0, 2mM DTT, 50mM NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers