missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADPRH Protein
A cDNA sequence encoding the ADPRH was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 141 |
---|---|
Name | ADPRH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | ADPRH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15905738
|
enQuireBio™
QP10971-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995728
|
enQuireBio™
QP10971-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985728
|
enQuireBio™
QP10971-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
141 | |
Research Use Only | |
ADPRH | |
Human | |
His | |
The ADPRH solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 100mM NaCl. |
ADPRH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEKYVA AMVLSAAGDA LGYYNGKWEF LQDGEKIHRQ LAQLGGLDAL DVGRWRVSDD TVMHLATAEA LVEAGKAPKL TQLYYLLAKH YQDCMEDMDG RAPGGASVHN AMQLKPGKPN GWRIPFNSHE GGCGAAMRAM CIGLRFPHHS QLDTLIQVSI ESGRMTHHHP TGYLGALASA LFTAYAVNSR PPLQWGKGLM ELLPEAKKYI VQSGYFVEEN LQHWSYFQTK WENYLKLRGI LDGESAPTFP ESFGVKERDQ FYTSLSYSGW GGSSGHDAPM IAYDAVLAAG DSWKELAHRA FFHGGDSDST AAIAGCWWGV MYGFKGVSPS NYEKLEYRNRLEETARALYS LGSKEDTVIS L | |
Greater than 90% as determined by SDS-PAGE. |