missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human aFGF / FGF1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5339-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000791.1 | |
aFGF / FGF1 Protein | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
E. coli | |
MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D | |
Greater than 95% as determined by SDS-PAGE. |
2246 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 75 mM sodium chloride, pH 7.5 |