missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AICDA Protein
A cDNA sequence encoding the AICDA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10985-20ug
Additional Details : Weight : 0.01000kg
Specifications
57379 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDSLLMNRRK FLYQFKNVRW AKGRRETYLC YVVKRRDSAT SFSLDFGYLR NKNGCHVELL FLRYISDWDL DPGRCYRVTW FTSWSPCYDC ARHVADFLRG NPNLSLRIFT ARLYFCEDRK AEPEGLRRLH RAGVQIAIMT FKDYFYCWNT FVENHERTFKAWEGLHENSV RLSRQLRRIL LPLYEVDDLR DAFRTLGL | |
Greater than 85.0% as determined by SDS-PAGE. |
AICDA Protein | |
Research Use Only | |
AICDA | |
Human | |
His | |
The AICDA solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |