missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AK4 Protein
A cDNA sequence encoding the AK4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10991-5ug
Additional Details : Weight : 0.01000kg
Spécification
205 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASKLLRAVI LGPPGSGKGT VCQRIAQNFG LQHLSSGHFL RENIKASTEV GEMAKQYIEK SLLVPDHVIT RLMMSELENR RGQHWLLDGF PRTLGQAEAL DKICEVDLVI SLNIPFETLK DRLSRRWIHP PSGRVYNLDF NPPHVHGIDD VTGEPLVQQE DDKPEAVAAR LRQYKDVAKP VIELYKSRGV LHQFSGTETN KIWPYVYTLF SNKITPIQSK EAY | |
Greater than 90% as determined by SDS-PAGE. |
AK4 Protein | |
Research Use Only | |
AK4 | |
Human | |
His | |
AK4 Human solution (1 mg/1ml) containing 20mM Tris-HCl pH 8.0, 2mM DTT, 0.1M NaCl, and 10% glycerol. |