missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKR1A1 Protein

A cDNA sequence encoding the AKR1A1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10994-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955808

  • 131.00 € / 5µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AKR1A1 Protein
Research Use Only
Recombinant Protein
E. coli
MAASCVLLHT GQKMPLIGLG TWKSEPGQVK AAVKYALSVG YRHIDCAAIY GNEPEIGEAL KEDVGPGKAV PREELFVTSK LWNTKHHPED VEPALRKTLA DLQLEYLDLY LMHWPYAFER GDNPFPKNAD GTICYDSTHY KETWKALEAL VAKGLVQALG LSNFNSRQID DILSVASVRP AVLQVECHPY LAQNELIAHC QARGLEVTAY SPLGSSDRAW RDPDEPVLLE PVVLALAEKY GRSPAQILL RWQVQRKVIC IPKSITPSRI LQNIKVFDFT FSPEEMKQLN ALNKNWRYIV PMLTVDGKRV PRDAGHPLYP FNDPY
Greater than 90% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
AKR1A1 solution containing 20mM Tris pH 8.0, 50mM NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers