missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKR1B1 Protein
A cDNA sequence encoding the AKR1B1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10511-1mg
Additional Details : Weight : 0.01000kg
Specifications
AKR1B1 Protein | |
Research Use Only | |
Specific activity is > 800pmol/min/ug, and is defined as the amount of enzyme that catalyze the reduction of 1.0 pmole DL-glyceraldehyde in the presence of NADPH per minute at pH7.0 at 37C. | |
Human | |
Untagged | |
The 1 mg/ml protein solution contains 20mM Tris-HCl buffer pH 8.0, 10% glycerol, and 1mM DTT. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MASRLLLNNG AKMPILGLGT WKSPPGQVTE AVKVAIDVGY RHIDCAHVYQ NENEVGVAIQ EKLREQVVKR EELFIVSKLW CTYHEKGLVK GACQKTLSDL KLDYLDLYLI HWPTGFKPGK EFFPLDESGN VVPSDTNILD TWAAMEELVD EGLVKAIGIS NFNHLQVEMI LNKPGLKYKP AVNQIECHPY LTQEKLIQYC QSKGIVVTAY SPLGSPDRPW AKPEDPSLLE DPRIKAIAAK HNKTTAQVLI RFPMQRNLVV IPKSVTPERI AENFKVFDFE LSSQDMTTLL SYNRNWRVCA LLSCTSHKDY PFHEEF | |
Greater than 95.0% as determined by SDS-PAGE. |