missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKR1B1 Protein

A cDNA sequence encoding the AKR1B1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10511-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934098

  • 203.00 € / 50µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

AKR1B1 Protein
Research Use Only
Specific activity is > 800pmol/min/ug, and is defined as the amount of enzyme that catalyze the reduction of 1.0 pmole DL-glyceraldehyde in the presence of NADPH per minute at pH7.0 at 37C.
Human
Untagged
The 1 mg/ml protein solution contains 20mM Tris-HCl buffer pH 8.0, 10% glycerol, and 1mM DTT.
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MASRLLLNNG AKMPILGLGT WKSPPGQVTE AVKVAIDVGY RHIDCAHVYQ NENEVGVAIQ EKLREQVVKR EELFIVSKLW CTYHEKGLVK GACQKTLSDL KLDYLDLYLI HWPTGFKPGK EFFPLDESGN VVPSDTNILD TWAAMEELVD EGLVKAIGIS NFNHLQVEMI LNKPGLKYKP AVNQIECHPY LTQEKLIQYC QSKGIVVTAY SPLGSPDRPW AKPEDPSLLE DPRIKAIAAK HNKTTAQVLI RFPMQRNLVV IPKSVTPERI AENFKVFDFE LSSQDMTTLL SYNRNWRVCA LLSCTSHKDY PFHEEF
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers